missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Toluenesulfonyl)pyrrole, 98%

Product Code. 11422100
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422100 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11422100 Supplier Thermo Scientific Alfa Aesar Supplier No. H28641.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17639-64-4 | C11H11NO2S | 221.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17639-64-4
Molecular Formula C11H11NO2S
Molecular Weight (g/mol) 221.274
MDL Number MFCD00145014
InChI Key OXWIEWFMRVJGNY-UHFFFAOYSA-N
Synonym 1-tosylpyrrole, 1-tosyl-1h-pyrrole, 1-p-toluenesulfonyl pyrrole, n-tosylpyrrole, 1-4-methylphenyl sulfonyl-1h-pyrrole, 1-p-tolylsulfonyl pyrrole, 1-tosyl-pyrrole, 1-4-methylphenylsulfonyl pyrrole, 1-4-methylbenzenesulfonyl pyrrole, 1h-pyrrole, 1-4-methylphenyl sulfonyl
PubChem CID 640087
IUPAC Name 1-(4-methylphenyl)sulfonylpyrrole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N2C=CC=C2

Specifications

Melting Point 99°C to 103°C
Quantity 25 g
Formula Weight 221.28
Percent Purity 98%
Chemical Name or Material 1-(p-Toluenesulfonyl)pyrrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.