missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Toluenesulfonyl)indole, 95%

Product Code. 11466016
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466016 25 g 25g
11446016 1 g 1g
11456016 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466016 Supplier Thermo Scientific Alfa Aesar Supplier No. B22320.14
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31271-90-6 | C15H13NO2S | 271.33 g/mol

1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31271-90-6
Molecular Formula C15H13NO2S
Molecular Weight (g/mol) 271.33
MDL Number MFCD01165032
InChI Key JNRRPYFLDADLJW-UHFFFAOYSA-N
Synonym 1-tosyl-1h-indole, n-tosylindole, 1-p-toluenesulfonyl indole, 1-4-methylbenzenesulfonyl indole, n-p-toluenesulfonyl indole, 1-4-methylphenyl sulfonyl-1h-indole, 1-tosylindole, 1-p-tolylsulfonyl indole, 1-p-toluenesulphonyl indole, 4-methylphenyl sulfonyl indole
PubChem CID 10956616
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12

Specifications

Melting Point 84°C to 86°C
Quantity 25 g
Solubility Information Insoluble in water.
IUPAC Name 1-(4-methylphenyl)sulfonylindole
Formula Weight 271.33
Percent Purity 95%
Chemical Name or Material 1-(p-Toluenesulfonyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.