missing translation for 'onlineSavingsMsg'
Learn More

1-Oxo-1H-isochromene-4-carboxylic Acid, TRC

Product Code. 30428759
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428759 100 mg 100mg
30424959 50 mg 50mg
30413352 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428759 Supplier Toronto Research Chemicals Supplier No. TRCO991653100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15868-29-8
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.15
IUPAC Name 1-oxoisochromene-4-carboxylic acid
SMILES OC(=O)C1=COC(=O)c2ccccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H6O4/c11-9(12)8-5-14-10(13)7-4-2-1-3-6(7)8/h1-5H,(H,11,12)
Formula Weight 190.0266
Chemical Name or Material 1-Oxo-1H-isochromene-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.