Learn More
1-Octyne, 98%
Description
1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.
Solubility
Miscible with alcohol, ether and other organic solvents. Immiscible with water.
Notes
Incompatible with oxidizing agents. Keep away from heat, sparks, sources of ignition and flame.
Specifications
Specifications
| Melting Point | -60°C |
| Density | 0.746 |
| Boiling Point | 123°C to 126°C |
| Flash Point | 17°C (62°F) |
| Assay Percent Range | 98% |
| Linear Formula | CH3(CH2)5C≡CH |
| Refractive Index | 1.416 |
| Quantity | 10 g |
| UN Number | UN3295 |
| Beilstein | 1734494 |
| Show More |
Safety and Handling
- Danger
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H304-May be fatal if swallowed and enters airways.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P331-Do NOT induce vomiting.
- MIXTURE LIST-Contain: 1-Octyne
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.