missing translation for 'onlineSavingsMsg'
Learn More

1-Octylphosphonic dichloride, 97%

Product Code. 11359425
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359425 5 g 5g
11369425 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359425 Supplier Thermo Scientific Alfa Aesar Supplier No. L18587.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3095-94-1 | C8H17Cl2OP | 231.097 g/mol

1-Octylphosphonic dichloride is employed in the surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride and qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylphosphonic dichloride is employed in the surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride and qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3095-94-1
Molecular Formula C8H17Cl2OP
Molecular Weight (g/mol) 231.097
MDL Number MFCD00015813
InChI Key RTHCRCDXILKTCY-UHFFFAOYSA-N
Synonym octylphosphonic dichloride, 1-octylphosphonic dichloride, octylphosphonoyl dichloride, acmc-1afrz, octyl phosphonic dichloride, dichloro octyl phosphine oxide, phosphonic dichloride,p-octyl
PubChem CID 76541
IUPAC Name 1-dichlorophosphoryloctane
SMILES CCCCCCCCP(=O)(Cl)Cl

Specifications

Density 1.115
Boiling Point 143°C to 145°C (12 mmHg)
Refractive Index 1.4665
Quantity 5 g
UN Number UN3265
Beilstein 1931202
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 231.1
Percent Purity 97%
Chemical Name or Material 1-Octylphosphonic dichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.