missing translation for 'onlineSavingsMsg'
Learn More

1-Octyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7047338
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3158-26-7
C9H17NO
155.241
MFCD00013887
DYQFCTCUULUMTQ-UHFFFAOYSA-N
octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate
76616
1-isocyanatooctane
CCCCCCCCN=C=O
This item is not returnable. View return policy
3158-26-7
C9H17NO
155.241
MFCD00013887
DYQFCTCUULUMTQ-UHFFFAOYSA-N
octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate
76616
1-isocyanatooctane
CCCCCCCCN=C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.88
Boiling Point 200°C to 204°C
Flash Point 71°C (159°F)
Odor Pungent
Refractive Index 1.432
Quantity 1 g
UN Number UN2206
Sensitivity Moisture sensitive
Formula Weight 155.24
Percent Purity 97%
Chemical Name or Material 1-Octyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'