missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-one, 97%, stab. with 0.1% BHA

Product Code. 11387166
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387166 1 g 1g
11397166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387166 Supplier Thermo Scientific Alfa Aesar Supplier No. L07189.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4312-99-6 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4312-99-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00036558
InChI Key KLTVSWGXIAYTHO-UHFFFAOYSA-N
Synonym 1-octen-3-one, vinyl amyl ketone, amyl vinyl ketone, pentyl vinyl ketone, unii-7lt7z4q9xr, 1-octene-3-one, fema no. 3515, 7lt7z4q9xr, n-pentyl vinyl ketone, octen-3-one, 1
PubChem CID 61346
IUPAC Name oct-1-en-3-one
SMILES CCCCCC(=O)C=C

Specifications

Density 0.848
Boiling Point 56°C to 60°C (16 mmHg)
Flash Point 35°C (95°F)
Odor Characteristic
Refractive Index 1.436
Quantity 1 g
UN Number UN1224
Beilstein 1700905
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 1-Octen-3-one, stabilized with 0.1% BHA
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.