missing translation for 'onlineSavingsMsg'
Learn More

1-Octanethiol, TRC

Product Code. 30424323
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424323 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424323 Supplier Toronto Research Chemicals Supplier No. TRCO14856350MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Octanethiol
CAS 111-88-6
Molecular Formula C8 H18 S
Quantity 50 mg
Synonym 1-Mercaptooctane, 1-Octyl mercaptan, 1-Octylthiol, NSC 151955, NSC 41903, O 0025, Octyl mercaptan, Octylthiol, Thiokalcol 08, n-Octanethiol, n-Octyl mercaptan, n-Octylthiol
InChI Formula InChI=1S/C8H18S/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3
SMILES CCCCCCCCS
IUPAC Name octane-1-thiol
Molecular Weight (g/mol) 146.294
Formula Weight 146.1129
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.