missing translation for 'onlineSavingsMsg'
Learn More

1-Octanethiol, 97%

Product Code. 10743334 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10743334 2.5 kg Plastic Bottle 2.5kg
10685972 250 g Glass bottle 250g
10347570 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10743334 Supplier Thermo Scientific Acros Supplier No. 157480025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 111-88-6 | C8H18S | 146.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-88-6
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00004912
InChI Key KZCOBXFFBQJQHH-UHFFFAOYSA-N
Synonym 1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol
PubChem CID 8144
IUPAC Name octane-1-thiol
SMILES CCCCCCCCS

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8400g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic Bottle
Linear Formula CH3(CH2)7SH
Refractive Index 1.4525 to 1.4545 (20°C, 589nm)
Quantity 2.5 kg
Formula Weight 146.29
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1-Octanethiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Octanethiol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.