missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic acid sodium salt, monohydrate, 99.7%, For HPLC analysis, MP Biomedicals™

Product Code. 11422572 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422572 25 g 25g
17881371 5 g 5g
11432572 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422572 Supplier MP Biomedicals Supplier No. 0215177825
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Item Name 1-Octanesulfonic Acid, Sodium Salt, Monohydrate
Content And Storage Store at room temperature (15°C to 30°C)
Form Crystalline Powder
Quantity 25 g
Appearance White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.