missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octane-d17-thiol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Octanethiol-D17 |
| CAS | 226999-68-4 |
| Molecular Formula | C8 D17 H S |
| Quantity | 0.25 g |
| Synonym | 1-Octane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17-thiol (9 CI, ACI), 1-Octanethiol-d17, 1-Octane-d17-thiol, 1-Octanethiol-D17, 1-Mercaptooctane-d17, 1-Octyl-d17 mercaptan |
| InChI Formula | InChI=1 S/C8H18S/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])S |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecadeuteriooctane-1-thiol |
| Molecular Weight (g/mol) | 163.3983 |
| Formula Weight | 163.2196 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction