missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecylamine hydrochloride

Product Code. 11374088
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374088 25 g 25g
11364088 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374088 Supplier Thermo Scientific Alfa Aesar Supplier No. 041378.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1838-08-0 | C18H40ClN | 305.975 g/mol

1-Octadecylamine hydrochloride is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1838-08-0
Molecular Formula C18H40ClN
Molecular Weight (g/mol) 305.975
MDL Number MFCD00042018
InChI Key RNYJXPUAFDFIQJ-UHFFFAOYSA-N
Synonym octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl
PubChem CID 519552
IUPAC Name octadecan-1-amine;hydrochloride
SMILES CCCCCCCCCCCCCCCCCCN.Cl

Specifications

Melting Point 160°C
Quantity 25 g
Solubility Information Soluble in hot methanol.
Formula Weight 305.97
Physical Form Powder
Chemical Name or Material 1-Octadecylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.