missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecylamine, 95%

Product Code. 11348266
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348266 25 g 25g
11358266 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348266 Supplier Thermo Scientific Alfa Aesar Supplier No. L15458.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-30-1 | C18H39N | 269.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Octadecylamine is used as asphalt emulsifier, lubricant and dispersants. It acts as a cationic surfactant and find applications is hair rinse bases, wood preservatives, textile softeners, dyeing auxiliaries and pigment grinding aids. It is used in boiler as an additive, which prevents the corrosion arises due to steam. It is a basic building block and used as an intermediate in the preparation of their amine derivatives, ethoxylates and amides. Metal oxide nano crystals having controlled size and shape are prepared by the thermal decomposition of metal nitrates with octadecylamine.

Solubility
Soluble in chloroform, alcohol, ether, benzene and acetone.Insoluble in water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-30-1
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.52
MDL Number MFCD00008159
InChI Key REYJJPSVUYRZGE-UHFFFAOYSA-N
Synonym octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
PubChem CID 15793
ChEBI CHEBI:63866
IUPAC Name octadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCN

Specifications

Melting Point 53°C to 55°C
Density 0.862
Boiling Point 180°C to 183°C (5 mmHg)
Flash Point 148°C (298°F)
Linear Formula CH3(CH2)17NH2
Quantity 25 g
Beilstein 636111
Solubility Information Soluble in chloroform,alcohol,ether,benzene and acetone.Insoluble in water.
Formula Weight 269.52
Percent Purity 95%
Chemical Name or Material 1-Octadecylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.