missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanol, TRC

Product Code. 30137818
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137818 100 g 100g
30132982 5 g 5g
17987614 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137818 Supplier Toronto Research Chemicals Supplier No. TRCO235480100G
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-92-5
Molecular Formula C18 H38 O
Molecular Weight (g/mol) 270.49
Synonym Octadecan-1-ol, 1-Octadecanol (8CI, 9CI, ACI), 1-Hydroxyoctadecane, 1-Stearyl alcohol, Adol 62, Adol 64, Adol 68, Alfol 18, Alfol 18NF, Atalco S, Cachalot S 43, CO 1895, CO 1895F, CO 1897, CO 1898, Conol 1675, Conol 30F, Conol 30S, Conol 30SS, Crodacol S, Crodacol S 70, Crodacol S 95, Crodacol S 95 NF, Crodacol S 95EP, Elocol C 1899, Ginol 18, Hainol 18SS, Hyfatol 18-95, Hyfatol 18-98, Kalchol 8098, Kalcohl 80, Kalcohl 8098, Kalcohl 8099, Kalcol 8098, Kolliwax SA, Lanette 18, Lanette 18DEO, Lanette-S 3, Lanol S, Laurex 18, Lorol 28, Lorol C 18, n-Octadecanol, n-Octadecyl alcohol, Nacol 18-98, Nacol 1898, Nacol C 18, NSC 5379, Octadecanol, Octadecyl alcohol, Rofamol, Sipol S, Siponol S, Siponol SC, Speziol C 18 Pharma, Stearafine Pure, Stearic alcohol, Stearol, Stearyl alcohol, Stearyl alcohol NX, Steraffine, Tego Alkanol 18, ThaiOL 1898, VLTN 6
IUPAC Name octadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCCO

Specifications

Quantity 100 g
InChI Formula InChI=1S/C18H38O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3
Formula Weight 270.2923
Chemical Name or Material Stearyl alcohol (Octadecanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.