missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanol, 97%

Product Code. 11499872
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
This item is not returnable. View return policy

Product Code. 11499872

Brand: Thermo Scientific Alfa Aesar A12020.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-92-5 | C18H38O | 270.501 g/mol

1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-92-5
Molecular Formula C18H38O
Molecular Weight (g/mol) 270.501
MDL Number MFCD00002823
InChI Key GLDOVTGHNKAZLK-UHFFFAOYSA-N
Synonym stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
PubChem CID 8221
ChEBI CHEBI:32154
IUPAC Name octadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 56°C to 60°C
Boiling Point 210°C (15 mmHg)
Flash Point 174°C (345°F)
Assay Percent Range 97%
Quantity 2500 g
Beilstein 1362907
Merck Index 14,8805
Solubility Information Insoluble in water. Soluble in alcohol,acetone,ether,benzene,chloroform.
Formula Weight 270.5
Percent Purity 97%
Chemical Name or Material 1-Octadecanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.