missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanol, 95%, Thermo Scientific Chemicals

Catalog Number 10667534
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
112-92-5
C18H38O
270.5
MFCD00002823
GLDOVTGHNKAZLK-UHFFFAOYSA-N
stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
8221
CHEBI:32154
octadecan-1-ol
CCCCCCCCCCCCCCCCCCO
This item is not returnable. View return policy
112-92-5
C18H38O
270.5
MFCD00002823
GLDOVTGHNKAZLK-UHFFFAOYSA-N
stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
8221
CHEBI:32154
octadecan-1-ol
CCCCCCCCCCCCCCCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 1mg KOH/g max.
CAS Min % 94.0
CAS Max % 100.0
Melting Point 55.0°C to 58.0°C
Color White
Density 185g/mL
Boiling Point 210.0°C (15.0 mmHg)
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic bottle
Linear Formula CH3(CH2)17OH
Quantity 2.5 kg
Beilstein 01, 431
Merck Index 15, 8932
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and benzene
Formula Weight 270.5
Percent Purity 95%
Physical Form Flakes
Chemical Name or Material 1-Octadecanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'