missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 11367695
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367695 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11367695 Supplier Thermo Scientific Alfa Aesar Supplier No. L02406.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10147-41-8
Molecular Formula C18H37ClO2S
Molecular Weight (g/mol) 353.00
MDL Number MFCD00051521
InChI Key VDRWNKIQBYUKGD-UHFFFAOYSA-N
Synonym 1-octadecanesulfonyl chloride, octadecane-1-sulphonyl chloride, 1-octadecanesulphonyl chloride, acmc-20aoso, octadecanesulfonylchloride, octadecanesulfonyl chloride, 1-octadecanesulfonylchloride
PubChem CID 66281
IUPAC Name octadecane-1-sulfonyl chloride
SMILES CCCCCCCCCCCCCCCCCCS(Cl)(=O)=O

Specifications

Melting Point 56°C to 60°C
Quantity 25 g
UN Number UN3261
Beilstein 1794152
Sensitivity Moisture sensitive
Formula Weight 353.01
Percent Purity 95%
Chemical Name or Material 1-Octadecanesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.