missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanesulfonic acid, sodium salt, 90%

Product Code. 10730755 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10730755 10 g Glass bottle 10g
10698234 1 g Glass bottle 1g
10289143 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10730755 Supplier Thermo Scientific Acros Supplier No. 384910100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13893-34-0 | C18H37NaO3S | 356.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13893-34-0
Molecular Formula C18H37NaO3S
Molecular Weight (g/mol) 356.54
MDL Number MFCD00051526
InChI Key KBAFDSIZQYCDPK-UHFFFAOYSA-M
Synonym sodium 1-octadecanesulfonate, sodium octadecane-1-sulfonate, sodium stearylsulfonate, sodium octadecanesulfonate, 1-octadecanesulfonic acid sodium salt, sodium octadecane-1-sulphonate, 1-octadecanesulfonic acid, sodium salt, sodium sterylsulfonate, sodium oxadecanesulfonate, sodium n-octadecylsulfonate
PubChem CID 23668003
IUPAC Name sodium;octadecane-1-sulfonate
SMILES CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 163°C to 165°C
Color White
Elemental Analysis 88% min. (C, H)
Loss on Drying 5% max. (1 G, 105°C, 1 Hrs, 760mm)
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula CH3(CH2)17SO3Na
Quantity 10 g
Solubility Information Solubility in water: soluble
Formula Weight 356.54
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 1-Octadecanesulfonic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.