missing translation for 'onlineSavingsMsg'
Learn More

1-Octacosanol, 90+%, Thermo Scientific Chemicals

Product Code. 10441304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441304 1 g Glass Bottle 1g
10091564 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441304 Supplier Thermo Scientific Acros Supplier No. 258130010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 557-61-9
Molecular Formula C28H58O
Molecular Weight (g/mol) 410.77
MDL Number MFCD00044770
InChI Key CNNRPFQICPFDPO-UHFFFAOYSA-N
Synonym 1-octacosanol, octacosanol, montanyl alcohol, octacosyl alcohol, cluytyl alcohol, n-octacosanol, octacosanol-1, policosanol, octacosyl, polycosanol
PubChem CID 68406
ChEBI CHEBI:28243
IUPAC Name octacosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCO

Spécification

CAS Min % 90.0
CAS Max % 100.0
Melting Point 83.0°C
Color White
Boiling Point 200.0°C to 250.0°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)27OH
Quantity 1 g
Merck Index 15, 6833
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot hydrocarbons, hot dibutylether,, thf,,hot methylisobutylketone
Formula Weight 410.77
Percent Purity 90+%
Physical Form Crystalline Powder or Flaked Crystals
Chemical Name or Material 1-Octacosanol
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.