missing translation for 'onlineSavingsMsg'
Learn More

1-O-Methyl-2-deoxy-D-ribose, 90%

Product Code. 11406340
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11406340 1 g 1g
11416340 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11406340

Brand: Thermo Scientific Alfa Aesar H27181.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60134-26-1 | C6H12O4 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60134-26-1
Molecular Formula C6H12O4
Molecular Weight (g/mol) 148.16
MDL Number MFCD00134161
InChI Key NVGJZDFWPSOTHM-XCSHRFQENA-N
Synonym 1-o-methyl-2-deoxy-d-ribose, 2r,3s-2-hydroxymethyl-5-methoxytetrahydrofuran-3-ol, methyl 2-deoxy-d-ribofuranoside, 2r,3s-2-hydroxymethyl-5-methoxyoxolan-3-ol, methyl 2-deoxyribofuranoside, methyl-2-deoxy-d-erythro pentofuranoside, d-erythro-pentofuranoside, methyl 2-deoxy, pubchem9694, methyl 2-deoxy-d-erythro-pentofuranoside
PubChem CID 10154128
IUPAC Name (2R,3S)-2-(hydroxymethyl)-5-methoxyoxolan-3-ol
SMILES COC1C[C@H](O)[C@@H](CO)O1

Specifications

Density 1.224
Flash Point >110°C (230°F)
Refractive Index 1.469
Quantity 1 g
Optical Rotation +23° (c=1 in Water)
Formula Weight 148.17
Percent Purity 96%
Chemical Name or Material 1-O-Methyl-2-deoxy-D-ribose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.