missing translation for 'onlineSavingsMsg'
Learn More

1-Nonylamine, 97%

Product Code. p-7024045
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463737 25 g 25g
11473737 100 g 100g
11483737 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463737 Supplier Thermo Scientific Alfa Aesar Supplier No. A18209.14
Add to basket
This item is not returnable. View return policy

CAS: 112-20-9 | C9H22ClN | 179.73 g/mol

1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-20-9
Molecular Formula C9H22ClN
Molecular Weight (g/mol) 179.73
MDL Number MFCD00008249
InChI Key IMGHCWMUWVHYCO-UHFFFAOYSA-N
Synonym nonylamine, n-nonylamine, 1-aminononane, 1-nonanamine, 1-nonylamine, unii-7l7h2ht4ok, 7l7h2ht4ok, nonyl-amine, n-nonyl amine, amine c9
PubChem CID 16215
IUPAC Name nonan-1-amine
SMILES [Cl-].CCCCCCCCC[NH3+]

Specifications

Melting Point -1°C
Density 0.784
Boiling Point 200°C to 202°C
Flash Point 62°C (143°F)
Odor Amine-like
Refractive Index 1.433
Quantity 25 g
UN Number UN2735
Beilstein 1733633
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 143.27
Percent Purity ≥98%
Chemical Name or Material 1-Nonylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.