Learn More
1-Nonanol, 99%
Description
The primary use of nonanol is in the manufacture of artificial lemon oil. 1-Nonanol is mainly used in man-made rose essence, plasticizer, surfactant. 1-Nonanol was used as a reference material in the determination of airborne spore levels. 1-Nonanol inhibited the bacterial luciferase (BL) reaction in a dose-dependent manner.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The primary use of nonanol is in the manufacture of artificial lemon oil. 1-Nonanol is mainly used in man-made rose essence, plasticizer, surfactant. 1-Nonanol was used as a reference material in the determination of airborne spore levels. 1-Nonanol inhibited the bacterial luciferase (BL) reaction in a dose-dependent manner.
Solubility
Soluble in water (1 g/L) at 20°C, ether, ethanol, acetone, chloroform, and mineral oil.
Notes
Store at room temperature. Ensure good ventilation.
Specifications
Specifications
| Melting Point | -7°C |
| Density | 0.827 |
| Boiling Point | 214°C to 216°C |
| Flash Point | 75°C (167°F) |
| Odor | Pleasant |
| Linear Formula | CH3(CH2)8OH |
| Refractive Index | 1.433 |
| Quantity | 100 g |
| UN Number | UN3082 |
| Beilstein | 969213 |
| Show More |
Safety and Handling
- Nonalol
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: 1-Nonanol
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.