missing translation for 'onlineSavingsMsg'
Learn More

1-Nitrosopyrrolidine-2-carboxylic Acid, TRC

Product Code. 30470046
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470046 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470046 Supplier Toronto Research Chemicals Supplier No. TRCN497885250MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso-DL-proline
CAS 2571-28-0
Molecular Formula C5 H8 N2 O3
Quantity 250 mg
Synonym 1-Nitrosopyrrolidine-2-carboxylic Acid, Proline','1-nitroso- (9CI','ACI), 1-Nitrosoproline (ACI), DL-Proline','1-nitroso- (ZCI), 1-Nitrosopyrrolidine-2-carboxylic acid, NSC 109546, N-Nitroso-DL-proline, 1-Nitroso-DL-proline
InChI Formula InChI=1S/C5H8N2O3/c8-5(9)4-2-1-3-7(4)6-10/h4H,1-3H2,(H,8,9)
SMILES OC(=O)C1CCCN1N=O
IUPAC Name 1-nitrosopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 144.129
Formula Weight 144.053
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.