Learn More
1-Nitroso-2-naphthol, 98%
Description
1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.
Specifications
Specifications
| Melting Point | 104°C to 108°C (decomposition) |
| Quantity | 250 g |
| UN Number | UN3077 |
| Beilstein | 776947 |
| Merck Index | 14,6641 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 173.17 |
| Percent Purity | 99% |
| Chemical Name or Material | 1-Nitroso-2-naphthol |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.