missing translation for 'onlineSavingsMsg'
Learn More

1-Nitroso-2-naphthol, 98%

Product Code. 11493324
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493324 50 g 50g
11483324 10 g 10g
11403334 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493324 Supplier Thermo Scientific Alfa Aesar Supplier No. A10622.18
  • ONLINE EXCLUSIVE PRICE
    £74.30
    £66.65 / 50g
    Save £7.65
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 131-91-9 | C10H7NO2 | 173.171 g/mol

1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-91-9
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00003884
InChI Key YXAOOTNFFAQIPZ-UHFFFAOYSA-N
Synonym 1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
PubChem CID 8580
IUPAC Name 1-nitrosonaphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2N=O)O

Specifications

Melting Point 104°C to 108°C (decomposition)
Quantity 50 g
UN Number UN3077
Beilstein 776947
Merck Index 14,6641
Solubility Information Insoluble in water.
Formula Weight 173.17
Percent Purity 99%
Chemical Name or Material 1-Nitroso-2-naphthol
compliance-icons
Product Identifier
  • 1-Nitroso-2-naphthol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-Nitroso-2-naphthol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.