missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate, pure, indicator grade
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | >300.0°C |
| Color | Yellow to Orange |
| Loss on Drying | 5% max. (1 g, 105°C) |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Quantity | 500 g |
| Beilstein | 11, II, 190 |
| Merck Index | 15, 6731 |
| Solubility Information | Solubility in water: slightly soluble in water |
| Formula Weight | 377.25 |
| Show More |
Safety and Handling
Product Identifier
- 1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?