missing translation for 'onlineSavingsMsg'
Learn More

1-Nitropropane, 98%

Product Code. 11414004
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
This item is not returnable. View return policy

Product Code. 11414004

Brand: Thermo Scientific Alfa Aesar A11975.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-03-2 | C3H7NO2 | 89.094 g/mol

The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

Solubility
Soluble in water. (1.4 g/L)

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-03-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007407
InChI Key JSZOAYXJRCEYSX-UHFFFAOYSA-N
Synonym propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
PubChem CID 7903
ChEBI CHEBI:76261
IUPAC Name 1-nitropropane
SMILES CCC[N+](=O)[O-]

Specifications

Melting Point -108°C
Density 0.998
Boiling Point 131°C to 132°C
Flash Point 34°C (93°F)
Odor Unpleasant
Linear Formula CH3CH2CH2NO2
Refractive Index 1.402
Quantity 250 g
UN Number UN2608
Beilstein 506236
Merck Index 14,6627
Solubility Information Soluble in water. (1.4g/L)
Formula Weight 89.09
Percent Purity 98%
Chemical Name or Material 1-Nitropropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Nitropropane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Nitropropane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.