Learn More
1-Nitropropane, 98%
Beskrivning
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.
Solubility
Soluble in water. (1.4 g/L)
Notes
Store in cool place. Keep away from oxidizing agents.
Specifikationer
Specifikationer
| Melting Point | -108°C |
| Density | 0.998 |
| Boiling Point | 131°C to 132°C |
| Flash Point | 34°C (93°F) |
| Odor | Unpleasant |
| Linear Formula | CH3CH2CH2NO2 |
| Refractive Index | 1.402 |
| Quantity | 500 g |
| UN Number | UN2608 |
| Beilstein | 506236 |
| Visa mer |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.