missing translation for 'onlineSavingsMsg'
Learn More

1-Nitroadamantane, TRC

Product Code. 30133060
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133060 10 g 10g
17919644 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133060 Supplier Toronto Research Chemicals Supplier No. TRCN49140010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7575-82-8
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.23
Synonym 1-Nitrotricyclo[3.3.1.13,7]decane, NSC130990, NSC235788
IUPAC Name 1-nitroadamantane
SMILES [O-][N+](=O)[C@@]12C[C@@H]3C[C@@H](C[C@@H](C3)C1)C2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H15NO2/c12-11(13)10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9H,1-6H2/t7-,8+,9-,10-
Formula Weight 181.1103
Chemical Name or Material 1-Nitroadamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.