missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-3,5-bis(trifluoromethyl)benzene, 96%

Product Code. 11422828
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422828 5 g 5g
11432828 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422828 Supplier Thermo Scientific Alfa Aesar Supplier No. A16797.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 328-75-6 | C8H3F6NO2 | 259.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-75-6
Molecular Formula C8H3F6NO2
Molecular Weight (g/mol) 259.11
MDL Number MFCD00000384
InChI Key GMUWJDVVXLBMEZ-UHFFFAOYSA-N
Synonym 1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl nitrobenzene, 1,3-bis trifluoromethyl-5-nitrobenzene, 3,5-di trifluoromethyl nitrobenzene, benzene, 1-nitro-3,5-bis trifluoromethyl, 3,5-bis-trifluoromethylnitrobenzene, 3,5-di trifluoromethylnitro benzene, 3,5-bis-trifluoromethyl nitrobenzene, 5-nitro-1,3-bis trifluoromethyl benzene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-5-nitroxylene
PubChem CID 67603
IUPAC Name 1-nitro-3,5-bis(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Density 1.535
Boiling Point 75°C to 76°C (11 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.427
Quantity 5 g
Beilstein 1998251
Formula Weight 259.11
Percent Purity ≥97%
Chemical Name or Material 1-Nitro-3,5-bis(trifluoromethyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.