missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-2-(n-octyloxy)benzene, 98%

Product Code. p-7023598
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11473557

Brand: Thermo Scientific Alfa Aesar A17320.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37682-29-4 | C14H21NO3 | 251.326 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37682-29-4
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.326
MDL Number MFCD00014693
InChI Key CXVOIIMJZFREMM-UHFFFAOYSA-N
Synonym 2-nitrophenyl octyl ether, 1-nitro-2-octyloxy benzene, octyl o-nitrophenyl ether, 2-nitrophenyl n-octyl ether, 2-octyloxy nitrobenzene, benzene, 1-nitro-2-octyloxy, 1-2-nitrophenoxy octane, 1-nitro-2-octyloxybenzene, 2-n-octyloxy nitrobenzene, npoe
PubChem CID 169952
IUPAC Name 1-nitro-2-octoxybenzene
SMILES CCCCCCCCOC1=CC=CC=C1[N+](=O)[O-]

Specifications

Density 1.04
Boiling Point 163°C to 165°C (2 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.511
Quantity 5 g
Beilstein 2529905
Formula Weight 251.32
Percent Purity 98%
Chemical Name or Material 1-Nitro-2-(n-octyloxy)benzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.