missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-2,5-bis(trifluoromethyl)benzene, 98%, Thermo Scientific™

Product Code. 15481369
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481369 5 g 5g
15491369 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481369 Supplier Thermo Scientific Alfa Aesar Supplier No. H64260.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-88-7
Molecular Formula C8H3F6NO2
Molecular Weight (g/mol) 259.107
MDL Number MFCD00236674
InChI Key BZKVUOHHNCMTLH-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
PubChem CID 2734048
IUPAC Name 2-nitro-1,4-bis(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F

Specifications

Color Yellow
Density 1.570 g/mL
Boiling Point 90°C (15 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.427
Quantity 5 g
Beilstein 2508751
Sensitivity Light sensitive
Formula Weight 259.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Nitro-2,5-bis(trifluoromethyl)benzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.