missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylamine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11385188
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385188 2 g 2g
11395188 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385188 Supplier Thermo Scientific Alfa Aesar Supplier No. 044088.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Applied as a synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-46-5
Molecular Formula C10H10ClN
Molecular Weight (g/mol) 179.647
MDL Number MFCD00036370
InChI Key FOKKJVHTXPJHEN-UHFFFAOYSA-N
Synonym 1-naphthylamine hydrochloride, 1-naphthalenamine, hydrochloride, naphthalen-1-amine hydrochloride, 1-naphthylammonium chloride, 1-naphthalenamine hydrochloride, alpha-naphthylamine hydrochloride, 1-amino-naphthalene hydrochloride, 1-aminonaphthalene hcl, 1-naphthalenamine, hydrochloride 1:1, 1-aminonaphthalene hydrochloride
PubChem CID 11094
IUPAC Name naphthalen-1-amine;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2N.Cl

Specifications

Color Red to White
Quantity 2 g
UN Number UN2811
Sensitivity Air & Light Sensitive
Merck Index 14,6398
Solubility Information Soluble in water, alcohol, ether
Formula Weight 179.65
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Naphthylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.