missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylamine-7-Sulfonic Acid, Extra Pure, SLR, Fisher Chemical

Product Code. 10030280 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 10030280

Brand: Fisher Chemical N/0910/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 119-28-8
Melting Point 500°C
Color Beige-Gray
Packaging Plastic powder jar
Molecular Formula C10H9NO3S
MDL Number 4032
Quantity 100 g
Synonym 8-amino-2-naphthalenesulfonic acid, 1,7-cleve's acid, 1-naphthylamine-7-sulfonic acid, 2-naphthalenesulfonic acid, 8-amino, cleve's theta-acid, 1-amino-7-sulfonaphthalene, 1-aminonaphthalene-7-sulfonic acid, 8-naphthylamine-2-sulfonic acid, 1-amino-7-naphthalenesulfonic acid, 1-amino-naphthalene-7-sulphonic acid
InChI Key QEZZCWMQXHXAFG-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C(C=C2)S(=O)(=O)O)C(=C1)N
IUPAC Name 8-aminonaphthalene-2-sulfonic acid
Molecular Weight (g/mol) 223.246
PubChem CID 8386
Formula Weight 223.25g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • 1-Naphthylamine-7-sulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.