missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylacetonitrile, 97%

Product Code. 11306966
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306966 5 g 5g
11316966 25 g 25g
11326966 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11306966 Supplier Thermo Scientific Alfa Aesar Supplier No. L06696.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132-75-2 | C12H9N | 167.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-75-2
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.211
MDL Number MFCD00004041
InChI Key OQRMWUNUKVUHQO-UHFFFAOYSA-N
Synonym 1-naphthylacetonitrile, 2-naphthalen-1-yl acetonitrile, 1-naphthyl acetonitrile, 1-naphthaleneacetonitrile, acetonitrile, 1-naphthyl, alpha-naphthylacetonitrile, alpha-naphthyl acetonitrile, 1-naphthylmethylcyanide, .alpha.-naphthylacetonitrile, 2-1-naphthyl acetonitrile
PubChem CID 8596
IUPAC Name 2-naphthalen-1-ylacetonitrile
SMILES C1=CC=C2C(=C1)C=CC=C2CC#N

Specifications

Melting Point 33°C to 35°C
Boiling Point 180°C to 182°C (9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6192
Quantity 5 g
UN Number UN3439
Beilstein 1101012
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 1-Naphthylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.