missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylacetic acid, 95%, may cont. up to 5% 2-isomer

Product Code. 11478923
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11478923

Brand: Thermo Scientific Alfa Aesar A11069.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 86-87-3 | C12H10O2 | 186.21 g/mol

1-Naphthylacetic acid is used as a plant growth regulator and for plant tissue culture. It is used for the vegetative propagation of plants from stem and leaf cutting, abscission promoter and substrate for auxin inducers of specific plant enzymes. Further, it stimulates root formation in cuttings of woody and herbaceous plants. It is also used to prevent premature flower and fruit drop. It is also employed to control regrowth of tree sprouts after trimminguse. In addition to this, it is used in ornaments and serves as an ingredient in many commercial plant rooting horticultural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthylacetic acid is used as a plant growth regulator and for plant tissue culture. It is used for the vegetative propagation of plants from stem and leaf cutting, abscission promoter and substrate for auxin inducers of specific plant enzymes. Further, it stimulates root formation in cuttings of woody and herbaceous plants. It is also used to prevent premature flower and fruit drop. It is also employed to control regrowth of tree sprouts after trimminguse. In addition to this, it is used in ornaments and serves as an ingredient in many commercial plant rooting horticultural products.

Solubility
Slightly soluble in water, ethanol, acetone, chloroform and ether.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 86-87-3
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004046
InChI Key PRPINYUDVPFIRX-UHFFFAOYSA-N
Synonym 1-naphthylacetic acid, 1-naphthaleneacetic acid, 2-naphthalen-1-yl acetic acid, naphthalene-1-acetic acid, transplantone, phyomone, planofix, 1-naphthalene acetic acid, fruitone n, stop-drop
PubChem CID 6862
ChEBI CHEBI:32918
IUPAC Name 2-naphthalen-1-ylacetic acid
SMILES OC(=O)CC1=C2C=CC=CC2=CC=C1

Especificaciones

Melting Point 126°C to 133°C
Quantity 1000 g
Beilstein 1308415
Merck Index 14,6371
Solubility Information Slightly soluble in water,ethanol,acetone,chloroform and ether.
Formula Weight 186.21
Percent Purity 96%
Concentration or Composition (by Analyte or Components) May contain up to 4% 2-isomer
Chemical Name or Material 1-Naphthylacetic acid

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado