missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylacetic acid, 95%

Product Code. 10086043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10086043 500 g Plastic bottle 500g
10377411 25 g Glass bottle 25g
10387411 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10086043 Supplier Thermo Scientific Acros Supplier No. 349645000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-87-3 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-87-3
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004046
InChI Key PRPINYUDVPFIRX-UHFFFAOYSA-N
Synonym 1-naphthylacetic acid, 1-naphthaleneacetic acid, 2-naphthalen-1-yl acetic acid, naphthalene-1-acetic acid, transplantone, phyomone, planofix, 1-naphthalene acetic acid, fruitone n, stop-drop
PubChem CID 6862
ChEBI CHEBI:32918
IUPAC Name 2-naphthalen-1-ylacetic acid
SMILES OC(=O)CC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 126.0°C to 133.0°C
Color Beige or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 09, 666
Merck Index 15, 6456
Solubility Information Solubility in water: 0.38g/L (17°C). Other solubilities: soluble in aceton
Formula Weight 186.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1-Naphthylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1-Naphthylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.