missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl trifluoromethanesulfonate, 97%

Product Code. 15411238
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411238 1 g 1g
15431238 250 mg 250mg
15421238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411238 Supplier Thermo Scientific Alfa Aesar Supplier No. H55091.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Denne vare kan ikke returneres. Se returpolicy

CAS: 99747-74-7 | C11H7F3O3S | 276.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 99747-74-7
Molecular Formula C11H7F3O3S
Molecular Weight (g/mol) 276.229
MDL Number MFCD00192340
InChI Key WQWUQDVFRYMMCY-UHFFFAOYSA-N
Synonym 1-naphthyl trifluoromethanesulfonate, 1-naphthyl triflate, 1-naphthyl-triflate, 1-naphthalenyl triflate, acmc-20aot0, 1-trifluoromethanesulfonyl naphthalene
PubChem CID 378884
IUPAC Name naphthalen-1-yl trifluoromethanesulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2OS(=O)(=O)C(F)(F)F

Tekniske data

Color Red
Density 1.405 g/mL
Boiling Point 97°C to 98°C (0.3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.52
Quantity 1 g
UN Number UN3265
Formula Weight 276.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Naphthyl trifluoromethanesulfonate
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.