missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl PP1, MedChemExpress

Product Code. 16458557
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16458557

Brand: Medchem Express HY13941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 1-Naphthyl PP1 (1-NA-PP 1) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.

1-Naphthyl PP1 (1-NA-PP 1) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.

Specifications

Chemical Name or Material 1-Naphthyl PP1
Physical Form Solid
CAS 221243-82-9
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H19N5
For Use With (Application) Cancer-Kinase/protease
Synonym 1-NA-PP 1
Solubility Information DMSO : 12.5 mg/mL (39.38 mM; Need ultrasonic)
SMILES CC(N1N=C(C2=C3C=CC=CC3=CC=C2)C4=C(N)N=CN=C41)(C)C
Molecular Weight (g/mol) 317.39
Formula Weight 317.39
Percent Purity 98.11%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.