missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isothiocyanate, 98%

Product Code. 15448959
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448959 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15448959 Supplier Thermo Scientific Alfa Aesar Supplier No. L12418.09
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 551-06-4 | C11H7NS | 185.244 g/mol

1-Naphthyl isothiocyanate is a tool for the study of liver damage which causes bile stasis and hyperbilirubinemia acutely and bile duct hyperplasia and biliary cirrhosis chronically, with changes in hepatocyte function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl isothiocyanate is a tool for the study of liver damage which causes bile stasis and hyperbilirubinemia acutely and bile duct hyperplasia and biliary cirrhosis chronically, with changes in hepatocyte function.

So
TRUSTED_SUSTAINABILITY

Specifications

CAS 551-06-4
Melting Point 54°C to 57°C
Odor Irritating
Molecular Formula C11H7NS
MDL Number MFCD00003882
Quantity 10 g
UN Number UN2923
Beilstein 637868
Sensitivity Moisture sensitive
Merck Index 14,6411
Synonym 1-naphthyl isothiocyanate, 1-naphthylisothiocyanate, anit, kesscocide, alpha-naphthyl isothiocyanate, naphthalene, 1-isothiocyanato, naphthalene, isothiocyanato, isothiocyanic acid, 1-naphthyl ester, 1-naftylisothiokyanat, isothiocyanic acid 1-naphthyl ester
Solubility Information Hydrolyzes in water.
InChI Key JBDOSUUXMYMWQH-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=S
IUPAC Name 1-isothiocyanatonaphthalene
Molecular Weight (g/mol) 185.244
PubChem CID 11080
ChEBI CHEBI:35455
Formula Weight 185.25
Percent Purity 98%
Chemical Name or Material 1-Naphthyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.