missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isocyanate, 98%

Product Code. 11463417
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463417 25 g 25g
11414160 1 g 1g
11453417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463417 Supplier Thermo Scientific Alfa Aesar Supplier No. A17200.14
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 86-84-0 | C11H7NO | 169.183 g/mol

1-Naphthyl isocyanate is used in the preparation of cationic aromatic urethane. It is useful for the study of cholangiolitic hepatotoxicity in laboratory animals. It is also employed as separate enantiomers of beta-blockers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl isocyanate is used in the preparation of cationic aromatic urethane. It is useful for the study of cholangiolitic hepatotoxicity in laboratory animals. It is also employed as separate enantiomers of beta-blockers.

Solubility
Miscible with alcohol, chloroform and diethyl ether.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-84-0
Molecular Formula C11H7NO
MDL Number MFCD00003881
InChI Key BDQNKCYCTYYMAA-UHFFFAOYSA-N
Synonym 1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
PubChem CID 66589
IUPAC Name 1-isocyanatonaphthalene

Specifications

Melting Point 4°C
Density 1.177
Boiling Point 269°C
Flash Point 135°C (275°F)
Odor Irritating
Refractive Index 1.6335
Quantity 25 g
UN Number UN2206
Beilstein 742779
Sensitivity Moisture sensitive
Merck Index 14,6410
Solubility Information Miscible with alcohol,chloroform and diethyl ether.
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=O
Molecular Weight (g/mol) 169.183
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material 1-Naphthyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.