missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isocyanate, 99%

Product Code. p-2695886 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10444311

Brand: Thermo Scientific Acros 415410050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-84-0 | C11H7NO | 169.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-84-0
InChI Key BDQNKCYCTYYMAA-UHFFFAOYSA-N
Synonym 1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
PubChem CID 66589
IUPAC Name 1-isocyanatonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 4°C
Density 1.1800g/mL
Boiling Point 269°C to 270°C
Flash Point 135°C
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C11H7NO
Refractive Index 1.6324 to 1.6344
Quantity 5 g
Merck Index 15, 6492
Specific Gravity 1.18
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene,chloroform,ether,decomposes in alcohols
Molecular Weight (g/mol) 169.18
Formula Weight 169.18
Percent Purity 99%
Chemical Name or Material 1-Naphthyl isocyanate
Show More Show Less
compliance-icons
Product Identifier
  • 1-Naphthyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.