missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl acetate, 99%

Product Code. 11438087
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438087 50 g 50g
11448087 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438087 Supplier Thermo Scientific Alfa Aesar Supplier No. A17482.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 830-81-9 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-81-9
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00003922
InChI Key VGKONPUVOVVNSU-UHFFFAOYSA-N
Synonym 1-naphthyl acetate, 1-acetoxynaphthalene, alpha-naphthyl acetate, 1-naphthol, acetate, a-naphthyl acetate, naphthyl acetate, naphthalene, 1-acetoxy, 1-naphthalenol, acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate
PubChem CID 13247
IUPAC Name naphthalen-1-yl acetate
SMILES CC(=O)OC1=CC=CC2=CC=CC=C21

Specifications

Melting Point 43°C to 46°C
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 2046403
Sensitivity Moisture sensitive
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 1-Naphthyl acetate
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.