missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoxyacetic acid, 97%, Thermo Scientific™

Product Code. 10347561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10347561 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10347561 Supplier Thermo Scientific Acros Supplier No. 338920250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2976-75-2
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H10O3
Linear Formula C10H7OCH2COOH
MDL Number MFCD00003927
Quantity 25g
Synonym 1-naphthoxyacetic acid, 1-naphthyloxy acetic acid, 2-naphthalen-1-yloxy acetic acid, 1-naphthyloxyacetic acid, naphthyloxyacetic acid, 2-1-naphthyloxy acetic acid, unii-9qi2464poa, 1-naphthoxy acetic acid, naphthalen-1-yloxy acetic acid, acetic acid, 1-naphthalenyloxy
InChI Key GHRYSOFWKRRLMI-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2OCC(=O)O
IUPAC Name 2-naphthalen-1-yloxyacetic acid
Molecular Weight (g/mol) 202.209
PubChem CID 76313
ChEBI CHEBI:44588
Formula Weight 202.21
Percent Purity ≥96.0%
Chemical Name or Material 1-Naphthoxyacetic acid, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.