missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, 99+%

Product Code. 10417330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10417330 500 g Glass bottle 500g
10675942 100 g Glass bottle 100g
10217303 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10417330 Supplier Thermo Scientific Acros Supplier No. 128195000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Naphthol, 99+%, C10H8O, CAS Number-90-15-3, 1-hydroxynaphthalene, 1-naphthalenol, 1-naphthol, alpha-naphthol, basf ursol ern, fouramine ern, fourrine ern, furro er, naphthol, tertral ern, 100g, 06, 596, 278.0 deg.C to 280.0 deg.C, 100.0, 99.0, CHEBI:10319, Beige-Gray to Brown, 201-969-4, 125 deg.C | CAS: 90-15-3 | C10H8O | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-15-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.17
MDL Number MFCD00003930
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
Synonym 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
PubChem CID 7005
ChEBI CHEBI:10319
IUPAC Name naphthalen-1-ol
SMILES C1=CC=C2C(=C1)C=CC=C2O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 95.0°C to 97.0°C
Color Beige-Gray to Brown
Boiling Point 278.0°C to 280.0°C
Flash Point 125°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C10H7OH
Quantity 500 g
Beilstein 06, 596
Merck Index 15, 6468
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in benzene,alcohol and ether,freely soluble in chloroform,alkali hydroxide,solutions
Formula Weight 144.17
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material 1-Naphthol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Naphthol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.