missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoic hydrazide, 98+%

Product Code. 11362107
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11362107

Brand: Thermo Scientific Alfa Aesar L07942.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 43038-45-5 | C11H10N2O | 186.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43038-45-5
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD00016804
InChI Key VMFUMDXVTKTZQY-UHFFFAOYSA-N
Synonym 1-naphthohydrazide, 1-naphthhydrazide, 1-naphthalenecarbohydrazide, 1-naphthalenecarboxylic acid, hydrazide, 1-naphthoic hydrazide, .alpha.-naphthoylhydrazine, naphthalene-1-carboxylic acid hydrazide, naphthalenecarbohydrazide, a-naphthoylhydrazine, 1-naphthohydrazide #
PubChem CID 72689
IUPAC Name naphthalene-1-carbohydrazide
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)NN

Specifications

Melting Point 166°C to 168°C
Quantity 5 g
Beilstein 2966714
Formula Weight 186.21
Percent Purity ≥98%
Chemical Name or Material 1-Naphthoic hydrazide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.