missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoic acid, 98%

Product Code. 15790799 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15790799 100 g Plastic bottle 100g
10081470 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15790799 Supplier Thermo Scientific Acros Supplier No. 128181000
  • ONLINE EXCLUSIVE PRICE
    £74.90
    £65.65 / 100g
    Save £9.25
    12% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 86-55-5 | C11H8O2 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-55-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.18
MDL Number MFCD00004007
InChI Key LNETULKMXZVUST-UHFFFAOYSA-N
Synonym 1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
PubChem CID 6847
ChEBI CHEBI:36466
IUPAC Name naphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 158.0°C to 162.0°C
Color Brown to Yellow
Boiling Point 300.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C10H7CO2H
Quantity 100 g
Beilstein 09, 647
Merck Index 15, 6466
Solubility Information Solubility in water: slightly soluble in hot water. Other solubilities: freely soluble in hot alcohol and ether
Formula Weight 172.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Naphthoic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.