missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenemethanol, 98%

Product Code. 10144390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144390 10 g Glass bottle 10g
10701271 2 g Glass bottle 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10144390 Supplier Thermo Scientific Acros Supplier No. 180110100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4780-79-4 | C11H10O | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4780-79-4
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.20
MDL Number MFCD00004044
InChI Key PBLNHHSDYFYZNC-UHFFFAOYSA-N
Synonym 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol
PubChem CID 20908
ChEBI CHEBI:38137
IUPAC Name naphthalen-1-ylmethanol
SMILES OCC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 59°C to 62°C
Color Yellow
Boiling Point 301°C (715.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C10H7CH2OH
Quantity 10 g
Beilstein 06, 667
Formula Weight 158.2
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Naphthalenemethanol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.