missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneboronic acid, 97%

Product Code. p-7025625
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464727 1 g 1g
11474727 5 g 5g
11484727 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464727 Supplier Thermo Scientific Alfa Aesar Supplier No. B21219.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13922-41-3 | C10H9BO2 | 171.99 g/mol

1-Naphthaleneboronic acid is a very useful building block for introduction of a naphthyl group through cross-coupling protocols. It is useful in the synthesis of polyaromatic hydrocarbons utilizing the Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthaleneboronic acid is a very useful building block for introduction of a naphthyl group through cross-coupling protocols. It is useful in the synthesis of polyaromatic hydrocarbons utilizing the Suzuki reaction.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13922-41-3
Molecular Formula C10H9BO2
Molecular Weight (g/mol) 171.99
MDL Number MFCD00019722
InChI Key HUMMCEUVDBVXTQ-UHFFFAOYSA-N
Synonym 1-naphthaleneboronic acid, 1-naphthylboronic acid, naphthalene-1-boronic acid, naphthalenyl-1-boronic acid, 1-naphthalene boronic acid, 1-naphthaleneboronicacid, 1-naphthyleneboronic acid, boronic acid, naphthalenyl, 1-borononaphthalene, naphthylboronic acid
PubChem CID 254532
IUPAC Name naphthalen-1-ylboronic acid
SMILES OB(O)C1=CC=CC2=CC=CC=C12

Specifications

Melting Point 204°C to 210°C
Quantity 1 g
Beilstein 2937504
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 171.99
Percent Purity 97%
Chemical Name or Material 1-Naphthaleneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.