missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneacetic acid, MedChemExpress

Product Code. 16479997
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479997 Supplier Medchem Express Supplier No. HY18570
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 1-Naphthaleneacetic acid (1-Naphthylacetic acid), a synthetic auxin, can promote plant growth. 1-Naphthaleneacetic acid is also an inhibitor of PLA2, with an IC50 of 13.16 μM.

1-Naphthaleneacetic acid (1-Naphthylacetic acid), a synthetic auxin, can promote plant growth. 1-Naphthaleneacetic acid is also an inhibitor of PLA2, with an IC50 of 13.16 μM.

Specifications

Chemical Name or Material 1-Naphthaleneacetic acid
CAS 86-87-3
Color White
Purity Grade Notes Research
Molecular Formula C12H10O2
Quantity 5 g
Synonym 1-Naphthylacetic acid
Solubility Information DMSO : ≥ 100 mg/mL (537.03 mM) ∣H2O : 0.1 mg/mL (0.54 mM; Need ultrasonic)
SMILES OC(CC1=CC=CC2=C1C=CC=C2)=O
Molecular Weight (g/mol) 186.21
Formula Weight 186.21
Percent Purity 98.05%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.